ContactUsLinkComponent
SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Catalog # 24983-331
undefined
SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Catalog # 24983-331
Supplier Number:  H42053-0001
Specifications
  • Volumen:
    0.35 cu. ft. (internal)
  • Descripción:
    Desiccator Cabinet
  • Dimensiones exteriores:
    25W×25D×25H cm (927/32×927/32×927/32")
  • Referencia:
    24983-331
  • Nº de proveedor:
    420530001
Specifications
About this item
Perforated shelves are movable to suit your dry storage needs in this desiccator cabinet that stacks up to 5 units high.
  • Polystyrene construction
  • Adjustable, perforated shelves
  • Door disappears beneath cabinet when open
This smoke-tinted polystyrene cabinet has two removable shelves that can be set at four heights.
The pull-down door slides out of the way into a compartment located underneath the cabinet.
Ordering information: Includes a small desiccant tray.