Synthware Kuderna-Danish Flasks, Kemtech America
500 ml
Catalog # 60002-256
Supplier: KEMTECH AMERICA INC
Supplier #: F984500
Jump to:
Product Details & Documents
Top outer [ST] joint size: 24/40.
- Heavy wall borosilicate construction
- Ground glass joint
- Kuderna-Danish style
Lower inner [ST] joint size: 19/22.
Specifications
- Capacity:500 ml