Orders LinkContactUsLinkComponent
Synthware Distillation Receivers, Dean Stark
20 ml, 325 mm, 95 mm, 24/40
  60002-172
 :  D452424
Synthware Distillation Receivers, Dean Stark
  60002-172
Synthware Distillation Receivers, Dean Stark
Synthware Distillation Receivers, Dean Stark
20 ml, 325 mm, 95 mm, 24/40
$5,191.68
Each (1UN)
For easy separation of water from reaction or distillation system.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Graduated
  • Teflon® stopcock
With 2 mm PTFE stopcock. Graduated.
  • Width:
    95 mm
  • Capacity:
    20 ml
  • Height:
    325 mm
  • ST Joint Size:
    24/40