KIMAX® Chromatography Columns with Reservoir, Straight Stopcock Assembly, Kimble Chase
Catalog # 89001-654
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Descripción:Chromatography Column, 11 I.D. mm
- Referencia:89001-654
- Nº de proveedor:17810-11300
Specifications
About this item
These 300 mm columns feature a 250 ml reservoir and a reinforced top.
Lower end is tooled for extra strength and a secure, leakproof fit with the all-PTFE stopcock, ensuring that liquid contacts only glass and PTFE. An O-ring maintains tight seal over a 0 to 100 °C (32 to 212 °F) temperature range. Stopcock is easily removed for access to column packing.