KIMAX® Chromatographic Columns, Kimble Chase
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificados
Acerca de este artículo
With removable stopcock of PTFE and replaceable glass tip.
Lower end of the column is tooled for extra strength and tight, leakproof fit. Glass tip is supplied.
Frequently Bought Together