DECON-AHOL® 70% Isopropyl Alcohol Solution, Veltek Associates
Solution features 70% isopropyl alcohol and 30% water and is ideal for decontamination of gloves, surfaces, carts, countertops, and process lines, and for many other applications. Solution is available in nonsterile, sterile, and sterile WFI formats. Nitrogen-propelled aerosol does not permit aspiration of the room air into reservoir. Prefiltered to 0.2µm.
- Prefiltered to 0.2 µm
- Completely traceable and validated for sterility and shelf life
- Sterile WFI solutions use USP Water for Injection, are gamma irradiated
- Solution available in nonsterile, sterile, and sterile WFI formats
- Nitrogen-propelled aerosol does not permit aspiration of the room air into reservoir
: Tested according to USP specifications and delivered with lot-specific Certificate of Irradiation, Certificate of Analysis, LAL Testing Analysis, and Sterility Report.
: Packed in a double bag
Frequently Bought Together