ContactUsLinkComponent
Spor-Klenz® Ready-To-Use Sterilant/Disinfectant, STERIS®
Spor-Klenz® Ready-To-Use Sterilant/Disinfectant, STERIS®
Catalog # 21899-864
undefined
Spor-Klenz® Ready-To-Use Sterilant/Disinfectant, STERIS®
Catalog # 21899-864
Supplier Number:  62525-08EC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Tamaño:
    3.2 L (0.85 gal.)
  • Referencia:
    21899-864
  • Empaquetado:
    Plastic bottle, 4/case
  • Nº de proveedor:
    652501
Specifications
About this item
This fast-acting, liquid cold sterilant/disinfectant requires no mixing or activation. Use wherever critically high-level microbial control is required. Ready-to-use format eliminates errors in dilution preparation. Low-toxicity profile adds to worker safety. Undiluted, the product is sporicidal in 51/2 hours and disinfects in 10 minutes. Accompanied by certificates of analysis, filtration, and sterility.
  • Ease of Use - Consistent dilution every time because no mixing or activation is required
  • Efficacy - Microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC and EN protocols
  • Safety - Low toxicity profile to support worker safety
  • Convenience - Excellent material compatibility allows use on most environmental surfaces
  • Flexibility - Versatility of use as a sterilant, sporicide, disinfectant or sanitizer
STERIS' Spor-Klenz® Ready to use cold sterilant is a fast acting, liquid cold sterilant/disinfectant, filtered to 0.22 µm and specifically formulated for use in the sterilization and disinfection of hard environmental surfaces in Pharmaceutical, Medical Device, Biotech and Cosmetic manufacturing facilities. This product is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic that provides fast, effective microbial control, including spores. All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD). Spor-Klenz® Ready to use sterilant offers a low toxicity profile, requires no mixing or activation and is reusable for up to 14 days which reduces waste. Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility.