Orders LinkContactUsLinkComponent
Plastic Surface Cleaner, Stoner®
Plastic Surface Cleaner, Stoner®
Aeorosol Can, 368 g (13 oz.)
Catalog # 89065-930
Supplier:   STONER INC.
Número de proveedor #:  A163
Plastic Surface Cleaner, Stoner®
Catalog # 89065-930
Supplier:  STONER INC.
Plastic Surface Cleaner, Stoner®
Plastic Surface Cleaner, Stoner®
Aeorosol Can, 368 g (13 oz.)
Price
$148.58
Log in Para disponibilidad
Each(1/Itm/Bx) Case de 12 (1/ Itm/Bx)
Product Details & Documents
Water-based cleaner removes dirt, oil, toner, carbon, fingerprints, fog, and lint from plastic surfaces on computers, printers, photocopiers, telephones, fax machines, and other equipment
It can also be used on glass, vinyl, or painted surfaces. A special foaming action holds the cleaner in place until wiped, so it will not drip or run. Cleaner dries fast, adds shine to surface, and will not leave streaks, haze, or film.
Documents
Especificaciones
  • Description:
    Aeorosol Can
  • Size:
    368 g (13 oz.)