ContactUsLinkComponent
Tergajet® Low-Foaming Powdered Detergents
Tergajet® Low-Foaming Powdered Detergents
Catalog # 80077-369
Supplier:  ALCONOX INC
undefined
Tergajet® Low-Foaming Powdered Detergents
Catalog # 80077-369
Supplier:  ALCONOX INC
Supplier Number:  2204
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Tamaño:
    9×1.8 kg (4 lbs.)
  • Referencia:
    80077-369
  • Empaquetado:
    Box
  • pH:
    11.5
  • Nº de proveedor:
    2204
Specifications
About this item
Tergajet® is a nonionic, low foaming phosphate free powdered cleaner for use in labware washers, manual and ultrasonic cleaning.
  • Passes inhibitory residue test Dilute 1:200
  • Concentrated
  • Phosphate-free, biodegradable and readily disposable
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Low foaming heavy duty detergent for high performance results
  • Free rinsing to give you reliable results and no interfering residues
Ideal for under-counter and floor standing lab washing machines. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues.
Used to remove soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents. Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for aluminum.
Used to clean water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning compliant.
Cleaning method: Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-inplace and manual cleaning if proper skin and eye protection is used.
Note: Corrosion testing is advisable.
Certifications: FDA certified.