Orders LinkContactUsLinkComponent
Corning® Coliform Water Test Sample Container, Corning
Corning® Coliform Water Test Sample Container, Corning
Supplier:   Corning
Corning® Coliform Water Test Sample Container, Corning
Supplier:  Corning
Certificates
Corning® Coliform Water Test Sample Container, Corning
Corning® Coliform Water Test Sample Container, Corning
2 Options Available Below
Product Details & Documents
These graduated, sterile, cylindrical vials made from PP are used in testing for the presence of coliform bacteria, microbiological contaminants in drinking water.
  • One-piece container with attached lid to reduce chance of contamination
  • Locking arrow ensures sterility
  • Leak tight when sealed properly
  • Tie-down to protect from accidental opening
  • With or without sodium thiosulfate tablet
Meets EPA requirements
Ordering information: Includes 100 containers per case. Comes with sample labels and instructions.
Product Family Options
Información del producto
  • Capacity
    Cap included
    Availability
    Price
  • 66175-105
    Coliform water test sample container with Thiosulfate tablet
    Coliform water test sample container with Thiosulfate tablet
    120.00 ml
    Yes
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $3,380.00
    $3,380.00
    Case (100UN)
       $3,380.00
    Supplier Number: 1700-100
  • 27614-453
    Coliform Water Test Sample Container without Thiosulfate Tablet
    Coliform Water Test Sample Container without Thiosulfate Tablet
    120.00 ml
    Yes
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $3,230.00
    $3,230.00
    Case (100UN)
       $3,230.00
    Supplier Number: 1705-100