MICRO-90® Concentrated Cleaning Solution, International Products
Proveedor: DUBOIS CHEMICALS INC
Certificados
Acerca de este artículo
A powerful, multi-purpose liquid for precision cleaning of glassware, filter membranes, laboratory equipment, electronics, metal parts, plastics, and other surfaces
- Mildly alkaline, aqueous cleaner lifts, disperses, emulsifies, sequesters, dissolves, suspends, and decomposes soils
- Concentrate mixes instantly with water
- Safe to use
- Cleaner is environmentally friendly, with no CFCs, ODCs, phosphates, or phenols
Highly-concentrated — a one liter container of cleaner makes over 12 gallons at 2% solution.
Best used in manual, immersion, and low-pressure or low-agitation systems. Ideal for use in ultrasonic cleaners.
Certificaciones: Cleaner is NSF-registered as an A1, general cleaner in and around food-processing areas and equipment.
Información de pedido: Validation methods are available upon request.
Especificaciones
- Catalog No:
- 21830-420
- 89210-138
- 89210-140
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Certificación ambiental:
- NSF A1
- NSF A1
- NSF A1
Especificaciones
Información relacionada
Certificados
Buscar por número de loteFocus Safety Volume 1 Issue 1 VWR® Focus Production Volume 3
Documentación del producto
M90, NSF
Detalles adicionales del producto
Frequently Bought Together