Orders LinkContactUsLinkComponent
VWR® TraceClean® 60 ml Vials
Quality-Assured® (QA) Vials, 60.00 ml, 27.5×140 mm (1¹⁄₁₆×5½"), Clear, Yes, 24-414
  89093-864
VWR® TraceClean® 60 ml Vials
  89093-864
VWR® TraceClean® 60 ml Vials
VWR® TraceClean® 60 ml Vials
Quality-Assured® (QA) Vials, 60.00 ml, 27.5×140 mm (1¹⁄₁₆×5½"), Clear, Yes, 24-414
$3,132.78
Case (72UN)
Type I clear borosilicate glass vials are designed for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Supplied with pen-top caps
  • Prepared to avoid volatile contamination
Vials are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis in every case. QA vials also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) vials do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Supplied with pen-top caps that feature ultrasonically bonded 3.1 mm (¹/₈") fluoropolymer resin/silicone septa, which aid in volatile organic analyses.
  • Cap included:
    Yes
  • Cap Size:
    24-414
  • Capacity:
    60.00 ml
  • Color:
    Clear
  • Description:
    Quality-Assured® (QA) Vials
  • Diameter ×H:
    27.5×140 mm (1¹⁄₁₆×5½")