VWR® Jars, PTFE
473 ml (16 oz.), 125 mm, 90 mm
89026-170
Isostatically molded, pure FDA approved PTFE molded stackable jars have thick wall construction and a smooth internal finish.
- Chemically resistant and inert
- Screw-on cap included
Suitable for IPA Method 1311 TCLP.
Caution: Do not use with cap sealed for chemical reactions or digestions. Loosen lid when thermally sterilizing.
- Cap Size:90 mm
- Capacity:473 ml (16 oz.)
- Height:125 mm