Orders LinkContactUsLinkComponent
VWR® TraceClean® 20 ml Vials
VWR® TraceClean® 20 ml Vials
VWR® TraceClean® 20 ml Vials
Supplier:  VWR International
Certificates
VWR® TraceClean® 20 ml Vials
VWR® TraceClean® 20 ml Vials
3 Options Available Below
Product Details & Documents
Type I clear borosilicate glass vials are for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Available with polypropylene solid-top or open-top caps
  • Prepared to avoid volatile contamination
They are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis in every case. QA vials also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) vials do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Vials are available with polypropylene solid-top or open-top caps. Solid-top caps feature fluoropolymer resin liners. Open-top caps feature ultrasonically bonded 3.1 mm (¹/₈") fluoropolymer resin/silicone septa and are for volatile organic analyses.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Cap included
    Cap Type
    Cap Size
    Availability
    Price
  • 89096-774
    Processed (PC) Vials
    Processed (PC) Vials
    20.00 ml
    27.5×57 mm
    Yes
    Solid top
    24-414
    Log in for availability
    Log in for availability
    Case (72UN)
    Case (72UN) - $1,724.16
    $1,724.16
    Case (72UN)
       $1,724.16
  • 89093-848
    Not Processed (NPC) Vials
    Not Processed (NPC) Vials
    20.00 ml
    27.5×57 mm
    Yes
    Open top
    24-414
    Log in for availability
    Log in for availability
    Case (72UN)
    Case (72UN) - $1,900.43
    $1,900.43
    Case (72UN)
       $1,900.43
  • 89093-830
    Not Processed (NPC) Vials
    Not Processed (NPC) Vials
    20.00 ml
    27.5×57 mm
    Yes
    Solid top
    24-414
    Log in for availability
    Log in for availability
    Case (72UN)
    Case (72UN) - $1,561.12
    $1,561.12
    Case (72UN)
       $1,561.12