Orders LinkContactUsLinkComponent
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
Certificates
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
1 Options Available Below
Product Details & Documents
Sturdy screw-thread vials have a conical interior bottom for maximum retrieval of content.
Vials can be used for small-scale reactions, centrifugation, storage, and shipping. They are ideal for use in the biochemical, pharmaceutical, and radiochemical industries. Fire-glazed bottom protects against glass particulates. Low-particulate packaging protects against contamination during transportation and storage. Clear vials are made from Wheaton 33* low extractable borosilicate glass to safeguard against a change in product pH caused by alkali, which can be extracted by ordinary glass. Amber vials are made from Wheaton 320 low extractable borosilicate glass. Caps are black, phenolic solid-top or open-top with fluoropolymer resin-faced 14B rubber liners.
Certifications: Clear vials conform to ASTM Type I, Class A, and USP Type I requirements. Amber vials conform to USP Type I requirements.
Ordering information: These vials do not have interior ground joints.
Product Family Options
Información del producto
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Type
    Cap Size
    Availability
    Price
  • Vials with caps
    5.00 ml
    20×65 mm
    Clear
    Yes
    Solid top
    20-400
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (12UN)
    Case (12UN) - $6,971.21
    $6,971.21
    Case (12UN)
       $6,971.21
    Supplier Number: W986259NG
  • Vials with caps
    10.00 ml
    24×72 mm
    Clear
    Yes
    Open top
    24-400
    Discontinued
    Supplier Number: W986290NG