Orders LinkContactUsLinkComponent
Nalgene® Racks, Polypropylene, Thermo Scientific
40, 13 - 16 mm, 285×115×96 mm (11×4.5×3.7")
Catalog # 60985-217
 :  5930-0020
Nalgene® Racks, Polypropylene, Thermo Scientific
Catalog # 60985-217
Nalgene® Racks, Polypropylene, Thermo Scientific
Nalgene® Racks, Polypropylene, Thermo Scientific
40, 13 - 16 mm, 285×115×96 mm (11×4.5×3.7")
Price
$2,400.00
Log in Para disponibilidad
Each (1UN) Case de 4 (1UN)
Product Details & Documents
Widely spaced holes and three-tiered design facilitate insertion and removal of tubes
Holes are keyed with molded numbers and letters for easy identification. Superior chemical resistance. Not autoclavable.
Especificaciones
  • No. of Holes:
    40
  • L×W×H:
    285×115×96 mm (11×4.5×3.7")
  • For Tube O.D.:
    13 - 16 mm