ContactUsLinkComponent
PYREX® Dean Stark Distallation Receiver, [ST] Joints, Corning
PYREX® Dean Stark Distallation Receiver, [ST] Joints, Corning
  89091-104
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Dean Stark Distallation Receiver, [ST] Joints, Corning
  89091-104
 :  Corning
 :  3602-10
Product available on GSA Advantage®
   Low Manufacturing Impact
  • Capacidad:
    10 ml
  • Altura:
    216 mm (8½")
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Certificación ambiental:
    I-RECs, ISO 14001 - Monterrey
  • Referencia:
    89091-104
  • Nº de proveedor:
    3602-10
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This 10 ml Dean Stark distilling receiver is graduated from 0 to 1 ml in 0.1 ml intervals, with a maximum scale error of 0.05 ml. The graduation intervals from 1.0 to 10.0 ml are 0.2 ml, with a maximum scale error of 0.1 ml.
 : ASTM E-123