Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Proveedor: Thermo Fisher Scientific
Certificados
Acerca de este artículo
These racks are ideal for applications using 36 tubes or less
Racks for 13mm and 16mm tubes hold 36 tubes; racks for 20mm and 25mm tubes hold 20 tubes; racks for 30mm tubes hold 9 tubes.
These racks are made of polyketone.
They are molded of ResMer® resin, which has excellent chemical resistance and can be immersed in a water bath. Racks feature excellent thermal properties which allow both autoclaving and freezing. Letters and numbers on the racks aid in tube identification.
Especificaciones
- Catalog No:
- 66024-016
- 66024-010
- 66023-845
- 66024-018
- 66024-020
- 66024-000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)