Corning® Cap Inserts for Cryogenic Vials with Closures, Polypropylene, Corning
Color-Coded Cap Inserts, White
Supplier: Corning
Número de proveedor #: 2015
Corning® Cap Inserts for Cryogenic Vials with Closures, Polypropylene, Corning
Catalog # 29442-544
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
The white polypropylene cap insert provide color-coding for easy sample identification.
- Inserts are packaed in resealable bags
- Non-sterile
Documents
Especificaciones
- Color:White
- Description:Color-Coded Cap Inserts
- Cat. No.:29442-544
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact