Insect Breeding Containers, Greiner Bio-One
Catálogo # 82050-328
Proveedor: GREINER BIO-ONE
Product available on GSA Advantage®
Especificaciones
- Tapa incluida:No
- Capacidad:68.00 ml
- Referencia:82050-328
- Diá. x Al:36×83 mm
- Nº de proveedor:217101
Especificaciones
Acerca de este artículo
These polystyrene breeding containers are specifically designed for the handling, storage, and transportation of living insect specimens.
- Hydrophobic
- Nonsterile
- Lighter and stronger than glass
The clear polystyrene tubes are lighter and stronger than glass, yet provide the same optical clarity. The hydrophobic nature of polystyrene prevents moisture accumulation, maintaining dry specimen storage.
White ceaprene foam plugs (sold separately) permit unrestricted gas exchange.