VWR® SoftCIDE® Extra-Mild Hand Soap
Liquid soap in pump bottle, 473 ml (16 oz.)
Supplier: VWR International
Jump to:
Product Details & Documents
Unique formulation helps replenish skin’s natural oils that are rinsed away by frequent washing.
- Anti-microbial strength
- Effective against broad spectrum of microorganisms
- pH balanced for skin
Soap has antimicrobial strength but is extra mild on hands. Contains 1% chloroxylenol (PCMX), a fast-acting, broad spectrum antimicrobial ingredient that is effective against gram-negative and gram-positive bacteria as well as yeast.
Documents
Especificaciones
- Capacity:473 ml (16 oz.)
- Description:Liquid soap in pump bottle