VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Catalog # 12985-078
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Dimensiones:17.8 x 17.8 cm (7 x 7")
- Referencia:12985-078
Specifications
About this item
Support stand base is constructed of cast iron with black enamel finish.
- Cast iron construction with black enamel
- U-shaped base
- Built-in support rod holder
- For use with VWR® Talon® lab frames
Base features three built-in support rod holders for additional support or to customize your stand. Locking knob accepts a 1.3 cm (1/2") diameter support rod (sold separately).