Orders LinkContactUsLinkComponent
Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
Supplier:   DICKSON COMPANY
Temperature Chart Recorders, Dickson®
Supplier:  DICKSON COMPANY
Certificates
Temperature Chart Recorders, Dickson®
4 Options Available Below
Product Details & Documents
Recorders are ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability. Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers. Cord-free, portable, and easy to use, recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall. They feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door, and durable, corrosion-resistant black ABS and polycarbonate enclosure. Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times. They are °C/°F switchable.
  • Cord-free, portable, and easy to use
  • Recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall
  • Feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door
  • Durable, corrosion-resistant black ABS and polycarbonate enclosure
  • Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times
  • °C/°F switchable
Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers.
Average response time: 11 min. to move 63% of full scale
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. CE marked. IP21 rated.
Ordering information: Each recorder comes with a AA alkaline battery and a red pen. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 61280-128
  • 97006-908
  • 97006-910
  • 97006-894
  • Accuracy:
  • Pen: ±2% of full scale
    Display: ±2°C (±3.6°F)
  • Pen: ±2% of full scale
    Display: ±2°C (±3.6°F)
  • Chart size:
  • 10.2 cm (4") dia.
  • 10.2 cm (4") dia.
  • Display:
  • 4.6H x 1.6W cm (113/16 x 5/8")
  • 4.6H x 1.6W cm (113/16 x 5/8")
  • Battery life:
  • 3 years
  • 3 years
  • Dimensions:
  • 13.2H x 13.2W x 6.1D cm (53/16 x 53/16 x 213/32")
  • 13.2H x 13.2W x 6.1D cm (53/16 x 53/16 x 213/32")
  • Weight:
  • 295 g (10.4 oz.)
  • 295 g (10.4 oz.)
Especificaciones
Product Family Options
Accessories
Información del producto
  • Temperature Range
    Availability
    Price
  • 61280-128
    7-Day Chart
    7-Day Chart
    –30...50 °C
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 60
    Paquete de 60 - $2,756.22
    $2,756.22
    Paquete de 60
       $2,756.22
    Supplier Number: C181
    Especificaciones:
    More
    Specifications
    Cat. No.
    61280-128
    Accuracy
    Chart size
    Display
    Battery life
    Dimensions
    Weight
  • 97006-894
    7-Day Chart
    7-Day Chart
    –22...122 °F
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 60
    Paquete de 60 - $2,756.22
    $2,756.22
    Paquete de 60
       $2,756.22
    Supplier Number: C206
    Especificaciones:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    97006-894
    Accuracy
    Chart size
    Display
    Battery life
    Dimensions
    Weight
    Small Business Enterprise
Charts
Información del producto
  • Temperature Range
    Availability
    Price
  • 97006-908
    Recorder with Digital Display
    Recorder with Digital Display
    –18...37 °C (0...100 °F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,704.46
    $13,704.46
    Each (1UN)
       $13,704.46
    Supplier Number: SL4100
    Especificaciones:
    More
    Specifications
    Cat. No.
    97006-908
    Accuracy
    Pen: ±2% of full scale
    Display: ±2°C (±3.6°F)
    Chart size
    10.2 cm (4") dia.
    Display
    4.6H x 1.6W cm (113/16 x 5/8")
    Battery life
    3 years
    Dimensions
    13.2H x 13.2W x 6.1D cm (53/16 x 53/16 x 213/32")
    Weight
    295 g (10.4 oz.)
  • 97006-910
    Recorder with Digital Display
    Recorder with Digital Display
    –30...50 °C (–22...122 °F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,704.46
    $13,704.46
    Each (1UN)
       $13,704.46
    Supplier Number: SL4350
    Especificaciones:
    More
    Specifications
    Cat. No.
    97006-910
    Accuracy
    Pen: ±2% of full scale
    Display: ±2°C (±3.6°F)
    Chart size
    10.2 cm (4") dia.
    Display
    4.6H x 1.6W cm (113/16 x 5/8")
    Battery life
    3 years
    Dimensions
    13.2H x 13.2W x 6.1D cm (53/16 x 53/16 x 213/32")
    Weight
    295 g (10.4 oz.)
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in Para disponibilidad
    Paquete de 6 - $2,985.88