VWR® Talon® Hook Connectors
Hook Connector, Nickel-Plated Zinc Finish
Catalog # 60079-166
Supplier: VWR International
Jump to:
Product Details & Documents
Hook connectors allow one-handed assembly of two components with one adjustment screw.
- Simple
- Versatile
- Easy to use
Maximum grip size: 13 mm (1/2").
Documents
VWR Laboratory Equipments
Product Literature
Specifications
- Description:Hook Connector, Nickel-Plated Zinc Finish