Lyophilization-Style Stoppers, Rubber, Wheaton
Mouth with ID×OD 13×20 mm, Stopper, 3-leg lyophilization, Gray, Chlorobutyl
: W224100-202
Wheaton Lyophilization Stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability
Stoppers are autoclavable.
- Color:Gray
- Description:Stopper, 3-leg lyophilization
- Material:Chlorobutyl
- Fits:Mouth with ID×OD 13×20 mm