Unisex MetaGuard™ Lab Coats, White Swan/Meta
Proveedor: WHITE SWAN META
Certificados
Acerca de este artículo
Full-length lab coat is fluid-resistant and antistatic.
Made of 99% air-textured polyester/1% carbon fiber fabric, this 102cm (40") long coat has a deep back vent and back pleats for freedom of movement. With knit cuffs and open neck.
Especificaciones
- Catalog No:
- 10843-282
- 10828-554
- 10828-552
- Aplicaciones de vestimenta:
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Género:
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Tela:
- 99% air-textured polyester / 1% carbon fiber
- 99% air-textured polyester / 1% carbon fiber
- 99% air-textured polyester / 1% carbon fiber