Orders LinkContactUsLinkComponent
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
   Opciones sostenibles disponibles
Supplier:   EPPENDORF BIO TOOLS
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
   Opciones sostenibles disponibles
Certificates
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
1 Options Available Below
Product Details & Documents
Eppendorf Tubes® with snap caps are an ideal choice when working with medium-sized sample volumes (0.5 to 5.0 ml). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25,000×g), and sample recovery.
  • Designed for ease of use and user-friendly handling during sample preparation and processing
  • Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions
  • Specifically designed to enhance sample recovery, preserving the integrity of samples
  • Available in Eppendorf Quality, PCR Clean, Biopur®, Sterile, and Forensic DNA Grade quality for tested purity
Explore the versatility of Eppendorf 5.0 ml tubes, available in both colorless and amber options. Tailor the selection with various certified purities, ensuring precise and reliable sample recoveries. Experience the flexibility these tubes provide, allowing selection of the ideal combination for specific research needs.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Snap cap
Material: PP
Diameter: 17 mm
 : Certificate of Quality/Conformity, Lot-specific certificates for the purity grades PCR clean, Biopur, Sterile®, Forensic DNA Grade can be downloaded from the Eppendorf website.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89429-306
  • 89429-308
  • 89429-312
  • 89429-310
  • Autoclavable:
  • When open 121 °C, 20 min
  • When open 121 °C, 20 min
  • When open 121 °C, 20 min
  • When open 121 °C, 20 min
  • Max. RCF:
  • 25000×g
  • 25000×g
  • 25000×g
  • 25000×g
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Operating temperature range:
  • −86...100 °C
  • −86...100 °C
  • −86...100 °C
  • −86...100 °C
  • Volume:
  • 5.0 ml
  • 5.0 ml
  • 5.0 ml
  • 5.0 ml
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Especificaciones
Product Family Options
Biopur® (Sterile)
Información del producto
  • Color
    Packaging
    Availability
    Price
  • Colourless
    50 tubes, individually wrapped
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,229.31
    $3,229.31
    Each (1UN)
       $3,229.31
    Supplier Number: 0030119479
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89429-312
    Autoclavable
    When open 121 °C, 20 min
    Max. RCF
    25000×g
    Environmentally Preferable
    Low Manufacturing Impact
    Operating temperature range
    −86...100 °C
    Volume
    5.0 ml
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact