PYREX® Allihn Condensers, Corning
Catálogo # 89091-494
Proveedor: Corning
Especificaciones
- ST Joint:24/40
- Environmentally Preferable:Low Manufacturing Impact
- Referencia:89091-494
- Jacket Length:400 mm (15¼")
- Nº de proveedor:2480-400
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
Especificaciones
Acerca de este artículo
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These widely used 5 bulb condensers have a greater surface area than the corresponding Liebig type and are suitable for use in vacuum distillations. They have an outer standard taper joint at the top and an inner standard taper joint at the bottom with a drip tip. The tubulations have an Ø of approximately 10 mm.