Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
Catálogo # 60820-140
Proveedor: GLOBE SCIENTIFIC INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Especificaciones
- Color:Natural
- Referencia:60820-140
- For Tubes:12 mm
- Nº de proveedor:118127C
Especificaciones
Acerca de este artículo
Polyethylene flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes
Closures fit 12mm tubes, and are available in a variety of colors to simplify identification and selection.
They feature two flexible flanges that press firmly against the inside of the tube wall, thereby providing better leak-resistance than standard plug caps.