Polypropylene Screw Caps, Linerless, Kimble Chase
Natural, 18-415
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 73805B-18415
Polypropylene Screw Caps, Linerless, Kimble Chase
Catalog # 66010-682
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Polypropylene caps feature one-piece construction with a seal achieved by a unique interior design.
Caps withstand repeated autoclaving when loosened or removed from tubes.
Certifications: Designed from ASTM Specification E982
Documents
Especificaciones
- Cap Size:18-415
- Color:Natural
- Cat. No.:66010-682