Orders LinkContactUsLinkComponent
VWR® Caps
VWR® Caps
VWR® Caps
Supplier:  VWR International
Certificates
VWR® Caps
4 Options Available Below
Product Details & Documents
Safe-T-Flex caps feature a unique, flexible, over-locking design that locks securely on the outside of the tube lip and can be removed with one hand.
  • Polyethylene construction
  • Liquid-tight seal
  • Color-coded for easy specimen identification
  • Cap top can be labeled
  • Self-locking caps hold firmly in any position
A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples
Caps fit all varieties of blood collecting and disposable culture tubes. They are color-coded for easy specimen identification and to help guard against cross-contamination. Top of cap can also be numbered or labelled with indelible ink. Self-locking caps hold firmly in any position and keep contents secure while travelling. Liquid-tight seal withstands centrifugation, agitation, refrigeration, and freezing. Manufactured from polyethylene.
This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus.
Documents
Product Family Options
For 12 mm Culture Tubes and 13 mm Blood Collecting Tubes
Información del producto
  • Color
    Availability
    Price
  • Lavender
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1,000
    Paquete de 1,000 - $1,100.32
    $1,100.32
    Log in Para disponibilidad
    Case de 10 (1,000,UN)
    Case de 10 (1,000,UN) - $1,199.35
    $1,199.35
    Paquete de 1,000
       $1,100.32
    Case de 10 (1,000,UN)
       $1,199.35
For 13 mm Culture Tubes
Información del producto
  • Color
    Availability
    Price
  • Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1,000
    Paquete de 1,000 - $1,218.54
    $1,218.54
    Paquete de 1,000
       $1,218.54
For 16 mm Blood Collecting and Culture Tubes
Información del producto
  • Color
    Availability
    Price
  • Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1,000
    Paquete de 1,000 - $1,100.32
    $1,100.32
    Log in Para disponibilidad
    Case de 10 (1,000,UN)
    Case de 10 (1,000,UN) - $1,199.35
    $1,199.35
    Paquete de 1,000
       $1,100.32
    Case de 10 (1,000,UN)
       $1,199.35
  • Red
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1,000
    Paquete de 1,000 - $1,100.32
    $1,100.32
    Log in Para disponibilidad
    Case de 10 (1,000,UN)
    Case de 10 (1,000,UN) - $1,199.35
    $1,199.35
    Paquete de 1,000
       $1,100.32
    Case de 10 (1,000,UN)
       $1,199.35