Orders LinkContactUsLinkComponent
Phenolic Screw Thread Closure, Kimble Chase
16 mm, 15-415
  89001-504
Phenolic Screw Thread Closure, Kimble Chase
  89001-504
Phenolic Screw Thread Closure, Kimble Chase
Phenolic Screw Thread Closure, Kimble Chase
16 mm, 15-415
$19,424.94
Case (300UN)
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
  • Cap Size:
    15-415
  • Height:
    16 mm