VWR® Plug-Type Caps, PE
Acerca de este artículo
Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.
- Disposable
- Polyethylene construction
- Unique fluted design
- Snugly grasps plastic or glass tubes
They feature a unique fluted design that grips plastic or glass tubes snugly.
Ordering information: The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.
3 Options Available Below
Product Details & Documents
Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.
- Disposable
- Polyethylene construction
- Unique fluted design
- Snugly grasps plastic or glass tubes
They feature a unique fluted design that grips plastic or glass tubes snugly.
Ordering information: The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.
Specifications for Product Family
Especificaciones
- Catalog No:
- 60819-003
- 60819-080
- 60819-069
- Environmentally Preferable:
- Sustainable Packaging|Low Manufacturing Impact
- Sustainable Packaging|Low Manufacturing Impact
- Sustainable Packaging|Low Manufacturing Impact
Especificaciones
Product Family Options
Información del producto
- ColorAvailabilityPrice
- NaturalPaquete de 1,000$1,171.10Case de 3 (1,000,UN)$1,276.49Especificaciones:
Sustainable Packaging|Low Manufacturing Impact More
Specifications
Cat. No.60819-003Environmentally PreferableSustainable Packaging|Low Manufacturing ImpactSustainable Packaging|Low Manufacturing Impact - GreenPaquete de 1,000$1,412.10Case de 3 (1,000,UN)$1,539.18Especificaciones:
Sustainable Packaging|Low Manufacturing Impact More
Specifications
Cat. No.60819-080Environmentally PreferableSustainable Packaging|Low Manufacturing ImpactSustainable Packaging|Low Manufacturing Impact - Especificaciones:
Sustainable Packaging|Low Manufacturing Impact More
Specifications
Cat. No.60819-069Environmentally PreferableSustainable Packaging|Low Manufacturing ImpactSustainable Packaging|Low Manufacturing Impact