VWR® Advanced Coveralls
Coveralls are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable. These low-linting disposable garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
- Significant fluid and particulate barrier
- Durable, lightweight fabric
- Excellent breathability and water vapor transmission rate
- Soft tactile feel
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back.
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Available with an elastic hood and attached boot covers designed to reduce the risk of slipping in wet or dry conditions. Coveralls without a hood have a tapered collar. Serged seams. These advanced protection coveralls are made of PP.
: 25/Case.
Coveralls are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable. These low-linting disposable garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
- Significant fluid and particulate barrier
- Durable, lightweight fabric
- Excellent breathability and water vapor transmission rate
- Soft tactile feel
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back.
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Available with an elastic hood and attached boot covers designed to reduce the risk of slipping in wet or dry conditions. Coveralls without a hood have a tapered collar. Serged seams. These advanced protection coveralls are made of PP.
: 25/Case.
- Catalog No:
- 414004-298
- 414004-297
- 414004-296
- 414004-293
- 414004-291
- 414004-292
- 414004-295
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Cleanroom class:
- 1000/ISO 6
- 1000/ISO 6
- 1000/ISO 6
- 1000/ISO 6
- 1000/ISO 6
- 1000/ISO 6
- 1000/ISO 6
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Air permeability:
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- Bacterial filtration efficiency:
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- Significate barrier
- Significate barrier
- Significate barrier
- Significate barrier
- Significate barrier
- Significate barrier
- Significate barrier
- Water vapor transmission rate:
- 777 g/m²/24 h
- 777 g/m²/24 h
- 777 g/m²/24 h
- 777 g/m²/24 h
- 777 g/m²/24 h
- 777 g/m²/24 h
- 777 g/m²/24 h
- Tensile strength:
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- 21.7 lbs./in (warp); 25.0 lbs./in (filling)
- Tearing strength:
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- 3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)
- Thickness:
- 0.21 mm
- 0.21 mm
- 0.21 mm
- 0.21 mm
- 0.21 mm
- 0.21 mm
- 0.21 mm
- Weight:
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
Product Family Options
Standard coverall
- ColorClothing SizeChest SizeSleevePackaging
- WhiteS24"33¹/₂"25/CaseCase (25UN)$1,416.17Paleta de 30 (25UN)$1,543.63
Specifications
Cat. No.414004-298Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - WhiteM24¹/₄"34"25/CaseCase (25UN)$1,429.08Paleta de 30 (25UN)$1,557.69
Specifications
Cat. No.414004-297Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - WhiteL25"34"25/CaseCase (25UN)$1,466.70Paleta de 30 (25UN)$1,598.71
Specifications
Cat. No.414004-296Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - WhiteXL27"36¹/₂"25/CaseCase (25UN)$1,482.81Paleta de 30 (25UN)$1,616.27
Specifications
Cat. No.414004-293Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - WhiteXXL29³/₄"37"25/CaseCase (25UN)$1,656.41Paleta de 24 (25UN)$1,805.48
Specifications
Cat. No.414004-291Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - White3XL32"38"25/CaseCase (25UN)$1,751.00Paleta de 24 (25UN)$1,908.59
Specifications
Cat. No.414004-292Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m² - White5XL33¹/₂"42"25/CaseCase (25UN)$1,761.22Paleta de 24 (25UN)$1,919.73
Specifications
Cat. No.414004-295Closure typeZip closureCleanroom class1000/ISO 6PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IIWater resistanceSignificate barrierWater vapor transmission rate777 g/m²/24 hTensile strength21.7 lbs./in (warp); 25.0 lbs./in (filling)Tearing strength3.4 lbs./in (lengthwise yarns); 4.8 lbs./in (widthwise yarns)Thickness0.21 mmWeight56 g/m²