NALGENE® Polypropylene Screw Closures, Thermo Scientific
Catalog # 16219-102
Supplier: Thermo Fisher Scientific
Specifications
- Tamaño de la tapa:43 mm
- Referencia:16219-102
- Nº de proveedor:712150-0430
Specifications
About this item
Linerless closures are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable. Available in natural and amber.