VWR® Drying Rack, Polystyrene
Drying Rack, 4¼", 25", 14¾", 18"
Supplier: VWR International
Jump to:
Product Details & Documents
High-impact polystyrene rack has holes to fit 72 pegs.
- Color: Light blue
Lightweight and versatile, it accommodates cylinders, flasks, and test tubes. Pegs snap out easily. With 7.6 cm (3") draining ledge.
Ordering information: Supplied with seventy-two 10.2 cm (4") pegs, mounting kit, fixing template, and drainage tube. Additional pegs for use with test tubes are also available.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Depth:4¼"
- Width:18"
- Description:Drying Rack
- Height:25"
- Length:14¾"