Nalgene® Peg Racks, Polypropylene, Thermo Scientific
For O.D. Tubes, 2½", 7", 5", White
Catalog # 60985-400
Supplier: Thermo Fisher Scientific
Número de proveedor #: 5977-0013
Nalgene® Peg Racks, Polypropylene, Thermo Scientific
Catalog # 60985-400
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Racks hold sample collection tubes, reaction tubes, and test tubes
Racks consist of an array of pegs spaced to hold test tubes upright and display the contents. Pegs are also excellent for holding inverted tubes for draining/drying.
Also hold and dry electrophoresis and chromatography plates. Durable, chemical-resistant, and autoclavable. Racks sink in water bath.
Especificaciones
- Width:5"
- Color:White
- Description:For O.D. Tubes
- Height:2½"
- Length:7"
- Cat. No.:60985-400
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact