Orders LinkContactUsLinkComponent
VWR® Cuvette Racks, PP
VWR® Cuvette Racks, PP
Non-Stackable Rack, 11.7 x 10.8 x 11.1 cm (45/8 x 41/4 x 43/8")
Catalog # 58020-104
Supplier:   VWR International
VWR® Cuvette Racks, PP
Catalog # 58020-104
Supplier:  VWR International
VWR® Cuvette Racks, PP
VWR® Cuvette Racks, PP
Non-Stackable Rack, 11.7 x 10.8 x 11.1 cm (45/8 x 41/4 x 43/8")
Price
$790.19
Log in Para disponibilidad
Each (1UN) Case de 6 (1UN)
Product Details & Documents
These two-tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.
  • Racks accommodate 12 cuvettes of 10 mm light path
  • Racks will not scratch quartz cuvettes
  • Rack 58020-105 features Stack Rack™ Interlocking end plates for secure stacking
  • Manufactured from polypropylene, and steam Autoclavable at 121°C (250°F)
Only the corners of the cuvettes touch the racks, minimizing chance for contamination or damage.
Especificaciones
  • Description:
    Non-Stackable Rack
  • L×W×H:
    11.7 x 10.8 x 11.1 cm (45/8 x 41/4 x 43/8")