SC-3 System Coveralls, WORKLON®
Reusable coveralls are designed for use in microelectronics, semiconductor, disk drive, laser, and other industrial ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanrooms
Coveralls have raglan sleeves, a zipper front with two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar and ankles. Three vertical snaps on the back of the legs accommodate an adjustable boot attachment. For boots, see 14227-884 series. A color-coded hang loop denotes size.
The Burlington C3™ high density fabric dissipates static charges and guards against airborne particles and lint.
Frequently Bought Together