Orders LinkContactUsLinkComponent
VWR® Cube Racks
VWR® Cube Racks
Assorted, 108×114×114 mm (41/4×41/2×41/2")
Catalog # 82010-746
Supplier:   VWR International
VWR® Cube Racks
Catalog # 82010-746
Supplier:  VWR International
VWR® Cube Racks
VWR® Cube Racks
Assorted, 108×114×114 mm (41/4×41/2×41/2")
Price
$1,037.32
Log in Para disponibilidad
Paquete de 5 Case de 12 (5UN)
Product Details & Documents
These polypropylene racks can slide together in multiple orientations to work with different-sized tubes at the same time.
  • Polypropylene fabrication
  • Autoclavable
Each cube rack has four sides that hold either four 50 mL tubes, ten 15 mL conical tubes, twelve 12x75mm or 12x100mm tubes, or sixteen 1.5–2.0 mL microcentrifuge tubes.
Ordering information: Assorted package includes one each of blue, green, pink, yellow, and orange.
Especificaciones
  • Color:
    Assorted
  • W×D×H:
    108×114×114 mm (41/4×41/2×41/2")