Liebig Condenser, Chemglass
Catálogo # 80067-398
Proveedor: CHEMGLASS INC.
Small Business Enterprise
Especificaciones
- Descripción:Liebig Condenser
- Referencia:80067-398
- Nº de proveedor:CG-1218-A-20
Especificaciones
Acerca de este artículo
Small scale Liebig condenser with a [ST] outer joint at the top and a lower inner drip tip joint
Hose connections have an O.D. of 10mm at the middle serration. [ST] joint: 14/20.