Liebig Condenser, Chemglass
80067-368
Small Business Enterprise
- Descripción:Liebig Condenser
- Referencia:80067-368
- Nº de proveedor:CG-1218-01
Condenser has a [ST] outer joint at the top and a lower inner drip tip joint
Hose connections have an O.D. of 10mm at the middle serration. [ST] joint: 24/40.