SP Bel-Art 'Wire' Lead Flask Weight with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These lead weights can be coiled into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath.
- Red vinyl coating
- Available in two different weights
- Chemical-resistant
- Flexible and versatile
They can be coiled around flasks, threaded through handles or looped around bottle necks.
Available in two weights, they are coated with chemical resistant red Vikem® vinyl. The 30 cm (12") length and flexibility make them very versatile.
Frequently Bought Together