Orders LinkContactUsLinkComponent
VWR® Lead Ring Flask Weights
VWR® Lead Ring Flask Weights
VWR® Lead Ring Flask Weights
Supplier:  VWR International
Certificates
VWR® Lead Ring Flask Weights
VWR® Lead Ring Flask Weights
4 Options Available Below
Product Details & Documents
Protect and stabilize expensive glassware with Vikem® vinyl-coated lead rings.
  • Stabilize glassware immersed in water baths
  • Reduce floating, breakage and chemical spills
  • Coated with bright red Vikem® vinyl
  • Chemical-resistant
  • Will not scratch flasks
  • Available in five diameter/weight combinations
Vikem® vinyl coated lead rings stabilize glassware immersed in water baths, thereby reducing floating, breakage, and chemical spills.
Documents
Product Family Options
Información del producto
  • Fits Flask
    I.D.
    O.D.
    Weight
    Availability
    Price
  • 125 - 500 ml
    43 mm (1.7")
    76 mm (3.0")
    200 g (0.5 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $804.76
    $804.76
    Each (1UN)
       $804.76
  • 1000 ml or Larger
    71 mm (2.8")
    114 mm (4.5")
    900 g (2 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,098.27
    $1,098.27
    Each (1UN)
       $1,098.27
  • 250 - 1000 ml
    51 mm (2.0")
    89 mm (3.5")
    500 g (1 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $847.60
    $847.60
    Each (1UN)
       $847.60
  • 500 - 2000 ml
    56 mm (2.2")
    102 mm (4.0")
    700 g (1.5 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $993.04
    $993.04
    Each (1UN)
       $993.04