ContactUsLinkComponent
Synthware Concentrator Tubes, Graduated, Kemtech America
Synthware Concentrator Tubes, Graduated, Kemtech America
Catalog # 60002-262
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Concentrator Tubes, Graduated, Kemtech America
Catalog # 60002-262
Supplier Number:  C291910
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Capacidad:
    10 ml
  • Altura:
    ~125 mm
  • Referencia:
    60002-262
  • Intervalo de graduación:
    0 – 1 ml in 0.1; 2 – 10 ml in 1.0 ml
  • Nº de proveedor:
    C291910
Specifications
About this item
Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups.
  • Heavy wall Borosilicate construction
  • Graduated
  • Ground glass joint
Tube can be stoppered for storage. [ST] joint size: 19/22.