ContactUsLinkComponent
Stainless Steel Dissecting Kits, Walter Stern
  25640-002
undefined
Stainless Steel Dissecting Kits, Walter Stern
  25640-002
 :  330-002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Descripción:
    Nine-Piece Kit
  • Referencia:
    25640-002
  • Nº de proveedor:
    330-002
Choose from six- or nine-piece kits
Six-piece kit includes bent dissecting needle, glass pipet with rubber bulb, 114mm dissecting scissors (straight sharp points), 150mm dissecting scalpel, 114mm dissecting forceps (straight fine serrated tips), and 150mm plastic ruler.
Nine-piece kit includes two 150mm dissecting scalpels, tenaculum, 114mm dissecting scissors (straight sharp points), blowpipe, hooks and chains, cartilage knife, 150mm plastic ruler, and 114mm dissecting forceps with straight fine serrated tips.
Both come with a vinyl case.