CarePac® Test Weight Sets, METTLER TOLEDO®
97007-304
- Referencia:97007-304
- Peso:2 g, 50 g
- Nº de proveedor:11123003
These sets enable routine testing of balances by using two weights at maximum and minimum loads
- Routine testing of balances
- Multiple configurations
Weights are available in both ASTM and OIML configurations and are specified to validate process tolerances up to 0.03%. Sets include ergonomic tweezers and cleaning cloths. Customized third weights are available for customer-specific testing, such as minimum weight determination. Specially-designed cases house both types of weights and all equipment. Additional space is provided to accommodate a third weight.