VWR® Advanced Hood
White, One size
Supplier: VWR International
Jump to:
Product Details & Documents
Hood is manufactured from a specially formulated, breathable microporous fabric that is exceptionally clean and durable.
- Significant fluid and particulate barrier
- Durable, lightweight, soft cloth-like fabric
- Excellent breathability and water vapor transmission rate
Garment allows full range of motion and vision, and exhibits excellent water vapor transmission to optimize breathability and user comfort
Garment features ties to ensure a secure and comfortable fit.
Hood is rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Provides resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Especificaciones
- Clothing Size:One size
- Color:White
- H₂O vapor transmission rate (avg.):777 g/m²/24 hrs.
- Water resistance:Pass
- Tensile strength:25 (Filling)
- Weight:56 g/m²
- Bacterial filtration efficiency:0.999
- Tearing strength:4.8 (Widthwise Yarns)
- Thickness:0.21 mm
- Cat. No.:414004-503
- Air permeability:0.06
- Garment application:Laboratory
- Synth. blood penetration resistance:Pass
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:777