PYREX® Flat-Bottom Evaporating Dishes, Corning
Supplier: Corning
PYREX® Flat-Bottom Evaporating Dishes, Corning
Supplier: Corning
Certificates
Acerca de este artículo
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Dishes with rugged beaded rims and standard medium depth.
3 Options Available Below
Product Details & Documents
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Dishes with rugged beaded rims and standard medium depth.
Documents
Specifications for Product Family
Especificaciones
- Catalog No:
- 25315-024
- 25315-068
- 470211-464
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Información del producto
- CapacityDiameter ×HAvailabilityPrice
- Supplier Number: 3180-80Especificaciones:Product available on GSA Advantage®
More
Specifications
Cat. No.25315-024Environmentally PreferableEnvironmental certificationProduct available on GSA Advantage® - Supplier Number: 3180-105Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.25315-068Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 170 ml90×50 mmEach (1UN)$607.00Case de 6 (1UN)$661.99Supplier Number: 3180-90Especificaciones:
More
Specifications
Cat. No.470211-464Environmentally PreferableEnvironmental certification