Orders LinkContactUsLinkComponent
PYREX® Crystallizing Dishes, Corning
PYREX® Crystallizing Dishes, Corning
Supplier:   Corning
PYREX® Crystallizing Dishes, Corning
Supplier:  Corning
Certificates
PYREX® Crystallizing Dishes, Corning
PYREX® Crystallizing Dishes, Corning
4 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Documents
Product Family Options
Product Information
  • Diameter ×H
    Availability
    Price
  • 70×50 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $3,440.00
    $3,440.00
    Log in for availability
    Case de 4 (6UN)
    Case de 4 (6UN) - $6,968.41
    $6,968.41
    Paquete de 6
       $3,440.00
    Case de 4 (6UN)
       $6,968.41
    Supplier Number: 3140-70
  • 125×65 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 4
    Paquete de 4 - $5,660.00
    $5,660.00
    Log in for availability
    Case de 3 (4UN)
    Case de 3 (4UN) - $6,282.08
    $6,282.08
    Paquete de 4
       $5,660.00
    Case de 3 (4UN)
       $6,282.08
    Supplier Number: 3140-125
  • 170×90 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 2
    Paquete de 2 - $4,540.00
    $4,540.00
    Log in for availability
    Case de 4 (2UN)
    Case de 4 (2UN) - $5,865.91
    $5,865.91
    Paquete de 2
       $4,540.00
    Case de 4 (2UN)
       $5,865.91
    Supplier Number: 3140-170
  • 190×100 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 2
    Paquete de 2 - $5,370.00
    $5,370.00
    Log in for availability
    Case de 3 (2UN)
    Case de 3 (2UN) - $5,958.35
    $5,958.35
    Paquete de 2
       $5,370.00
    Case de 3 (2UN)
       $5,958.35
    Supplier Number: 3140-190